Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25873425UL
Description
PDLIM7 Polyclonal specifically detects PDLIM7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PDLIM7 | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
1110003B01Rik, ENIGMAProtein enigma, LIM domain protein, LIM mineralization protein, LMP, LMP1, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7 | |
Rabbit | |
Affinity Purified | |
RUO | |
9260 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
PDLIM7 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GKTPVCHQCHKVIRGRYLVALGHAYHPEEFVCSQCGKVLEEGGFFEEKGAIFCPPCYDVRYAPSCAKCKKKITGEIMHALKMTWH | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction