Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDLIM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110094 Supplier Invitrogen™ Supplier No. PA5110094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPQKASAPAA DPPRYTFAPS VSLNKTARPF GAPPPADSAP QQNGQPLRPL VPDASKQRLM ENTEDWRPRP GTGQSRSFRI LAHLTGTEFM QDPDEEHLKK S.

The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDLIM7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pdlim7
Gene Accession No. Q3TJD7, Q9NR12
Gene Alias 1110003B01Rik; ENIGMA; enigma (LIM domain protein); enigma protein; enigma protein isoform 4; hypothetical protein MGC65907; LIM domain protein; LIM mineralization protein; LIM mineralization protein 2; Lim mineralization protein 3; Lim1; LMP; LMP1; LMP3; LOW QUALITY PROTEIN: PDZ and LIM domain protein 7; pdlim7; pdlim7 protein; PDZ and LIM domain 7; PDZ and LIM domain 7 (enigma); PDZ and LIM domain protein 7; Protein enigma; Unknown (protein for MGC:129104); zgc:65907; zgc:85607
Gene Symbols Pdlim7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-190 of human PDLIM7 (NP_998801.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67399, 9260
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.