Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25708225UL
Description
PDP2 Polyclonal specifically detects PDP2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PDP2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EC 3.1.3, EC 3.1.3.43, KIAA1348[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 2, mitochondrial, PDP 2, PDPC 2, PPM2C2, protein phosphatase 2C, magnesium-dependent, catalytic subunit 2, Pyruvate dehydrogenase phosphatase catalytic subunit 2, pyruvate dehydrogenase phosphatase isoenzyme 2, pyruvate dehydrogenase phosphatase, catalytic subunit 2, pyruvate dehyrogenase phosphatase catalytic subunit 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
PDP2 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VPPTLNSSPCGGFTLCKAYRHTSTEEDDFHLQLSPEQINEVLRAGETTHKILDLESRVPNS | |
25 μL | |
Protein Phosphatase | |
57546 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction