Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDPK1 Mouse anti-Human, Monkey, Mouse, Rat, Clone:7E4G11, Abnova™

Catalog No. 89333082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-082 Supplier Abnova Corporation Supplier No. MAB16717
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human PDPK1.

Specifications

Antigen PDPK1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7E4G11
Conjugate Unconjugated
Description 3-phosphoinositide dependent protein kinase-1
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry (1:50-1:200)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene PDPK1
Gene Alias MGC20087, MGC35290, PDK1, PRO0461
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 457-556 of human PDPK1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5170
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.