Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDRG1 Antibody (OTI4F12), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273320PEATT594
Description
PDRG1 Monoclonal specifically detects PDRG1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
PDRG1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
PDRG1 | |
Full length recombinant protein of human PDRG1 (NP_110442) produced in HEK293T cells. | |
0.1 mL | |
81572.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI4F12 | |
Flow Cytometry | |
C20orf126p53 and DNA damage regulated 1, chromosome 20 open reading frame 126, dJ310O13.3, p53 and DNA-damage regulated 1, PDRGp53 and DNA damage-regulated protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction