Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDRG1 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
SDP

Catalog No. 502544078
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4078 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4078 Supplier OriGene Supplier No. CF808878
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDRG1 Monoclonal antibody specifically detects PDRG1 in Human samples. It is validated for Western Blot

Specifications

Antigen PDRG1
Applications Western Blot
Classification Monoclonal
Clone OTI1F5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene EBLN2
Gene Accession No. Q6P2I7
Gene Alias 1110004D19Rik; AU020945; AV006589; C20orf126; p53 and DNA damage regulated 1; p53 and DNA damage-regulated protein 1; p53 and DNA-damage regulated 1; PDRG; PDRG1; RGD1305096
Host Species Mouse
Immunogen Full length human recombinant protein of human EBLN2 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55096
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.