Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDRG1 Polyclonal Antibody
PDRG1 Polyclonal Antibody
Supplier: Thermo Scientific PA561420
Description
PDRG1 Polyclonal Antibody for IHC

Specifications
PDRG1 | |
Polyclonal | |
Unconjugated | |
PDRG1 | |
1110004D19Rik; AU020945; AV006589; C20orf126; p53 and DNA damage regulated 1; p53 and DNA damage-regulated protein 1; p53 and DNA-damage regulated 1; PDRG; PDRG1; RGD1305096 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUG6 | |
PDRG1 | |
Recombinant protein corresponding to Human PDRG1. Recombinant protein control fragment (Product #RP-99296). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction