Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584599 Supplier Invitrogen™ Supplier No. PA584599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLSPEAERVL RYLVEVEELA EEVLADKRQI V Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 87%.

PDRG may play a role in chaperone-mediated protein folding (Potential).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDRG1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDRG1
Gene Accession No. Q9NUG6
Gene Alias 1110004D19Rik; AU020945; AV006589; C20orf126; p53 and DNA damage regulated 1; p53 and DNA damage-regulated protein 1; p53 and DNA-damage regulated 1; PDRG; PDRG1; RGD1305096
Gene Symbols PDRG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDRG1. Recombinant protein control fragment (Product #RP-107248).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81572
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.