Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDRG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539326
Description
Specifications
| PDRG1 | |
| Polyclonal | |
| Unconjugated | |
| PDRG1 | |
| C20orf126, PDRG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 296278, 68559, 81572 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NUG6, P59048, Q642A0 | |
| PDRG1 | |
| A synthetic peptide derived from the internal region of human PDRG1 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Non-human Primate, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction