Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDS5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557756 Supplier Invitrogen™ Supplier No. PA557756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRRPKSESQG NATKNDDLNK PINKGRKRAA VGQESPGGLE AGNAKAPKLQ DLAKKAAPAE RQIDLQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. NOL5A is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. The protein encoded by this gene is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDS5A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDS5A
Gene Accession No. A4L9P7, Q29RF7, Q6A026
Gene Alias 9030416H16Rik; Cell proliferation-inducing gene 54 protein; E230024D05Rik; KIAA0648; mKIAA0648; PDS5; PDS5 cohesin associated factor A; PDS5, regulator of cohesion maintenance, homolog A; PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae); PDS5A; PIG54; RGD1307094; SCC112; SCC-112; Sister chromatid cohesion protein 112; sister chromatid cohesion protein PDS5 homolog A
Gene Symbols PDS5A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDS5A. Recombinant protein control fragment (Product #RP-96638).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23244, 305343, 71521
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.