Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pds5B Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501557859
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7859 Supplier Bethyl Laboratories, Inc Supplier No. A300537A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

This gene encodes a protein that interacts with the conserved protein complex termed cohesion. The cohesion complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.

Specifications

Antigen Pds5B
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PDS5B
Gene Accession No. Q4VA53, Q9NTI5
Gene Alias APRIN, AS3, CG008, androgen induced inhibitor of proliferation, androgen-induced proliferation inhibitor, androgen-induced prostate proliferative shutoff-associated protein AS3, androgen-induced shutoff 3, sister chromatid cohesion protein PDS5 homolog B
Gene Symbols PDS5B
Host Species Rabbit
Immunogen Between 1200 and 1250
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100710, 23047
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.