Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDS5B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12605R

Description
This gene encodes a protein that interacts with the conserved protein complex termed cohesin. The cohesin complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.Specifications
PDS5B | |
Polyclonal | |
Unconjugated | |
PDS5B | |
AI646570; androgen induced inhibitor of proliferation; Androgen-induced proliferation inhibitor; androgen-induced prostate proliferative shutoff associated protein; androgen-induced prostate proliferative shutoff associated protein AS3; Androgen-induced prostate proliferative shutoff-associated protein AS3; androgen-induced shutoff 3; APRIN; AS3; AW212954; CG008; Kiaa0979; mKIAA0979; PDS5 cohesin associated factor B; PDS5, regulator of cohesion maintenance, homolog B; PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae); PDS5B; Sister chromatid cohesion protein PDS5 homolog B | |
Rabbit | |
Protein A | |
RUO | |
23047 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q9NTI5 | |
PDS5B | |
KLH conjugated synthetic peptide derived from human PDS5B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction