Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDSS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565340
Description
Immunogen sequence: SAAAEVRAQVH RRKGLDLSQI PYINLVKHLT SACPNVCRIS RFHHTTPDSK THSGEKYTDP FKLGWRDLKG LYEDIRKELL ISTSELK Highest antigen sequence identity to the following orthologs - mouse 53%, rat 57%.
PDSS1 is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. PDSS1 catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in PDSS1 gene are a cause of coenzyme Q10 deficiency.The protein encoded by this gene is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in this gene are a cause of coenzyme Q10 deficiency.
Specifications
PDSS1 | |
Polyclonal | |
Unconjugated | |
PDSS1 | |
2610203G20Rik; 2700031G06Rik; All-trans-decaprenyl-diphosphate synthase subunit 1; COenzyme Q (ubiquinone) biosynthesis family member (coq-1)-like; coenzyme Q1 homolog; COQ1; COQ10D2; decaprenyl diphdsphate synthase subunit1; decaprenyl diphosphate synthase subunit 1; decaprenyl pyrophosphate synthase subunit 1; decaprenyl pyrophosphate synthetase subunit 1; decaprenyl-diphosphate synthase subunit 1; DPS; Dps1; hDPS1; hypothetical protein LOC550349; LOW QUALITY PROTEIN: decaprenyl-diphosphate synthase subunit 1; mDLP1; MGC70953; mSPS1; PDSS1; polyprenyl pyrophosphate synthetase; prenyl (decaprenyl) diphosphate synthase, subunit 1; prenyl (solanesyl) diphosphate synthase, subunit 1; prenyl diphosphate synthase, subunit 1; RP13-16H11.3; solanesyl-diphosphate synthase subunit 1; SPS; Sps1; subunit 1 of decaprenyl diphosphate synthase; subunit 1 of solanesyl diphosphate synthase; TPRT; TPT; TPT 1; trans-prenyltransferase; trans-prenyltransferase (TPT); trans-prenyltransferase 1; wu:fc11f12; wu:fd05d05; zgc:112058 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23590 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T2R2 | |
PDSS1 | |
Recombinant Human PDSS1. Recombinant protein control fragment (Product #RP-104758). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction