Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDSS2 Antibody (OTI1D12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273323MFV500
Description
PDSS2 Monoclonal antibody specifically detects PDSS2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
PDSS2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
57107 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1D12 | |
Flow Cytometry | |
All-trans-decaprenyl-diphosphate synthase subunit 2, bA59I9.3, C6orf210decaprenyl-diphosphate synthase subunit 2, Candidate tumor suppressor protein, chromosome 6 open reading frame 210, Decaprenyl pyrophosphate synthase subunit 2, decaprenyl pyrophosphate synthetase subunit 2, DLP1hDLP1, EC 2.5.1.91, prenyl (decaprenyl) diphosphate synthase, subunit 2, subunit 2 of decaprenyl diphosphate synthase | |
Full length human recombinant protein of human PDSS2(NP_065114) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction