Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDSS2 Monoclonal Antibody (OTI1D10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503936

Description
The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.Specifications
PDSS2 | |
Monoclonal | |
0.8 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q86YH6 | |
PDSS2 | |
Full length human recombit protein of human PDSS2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D10 | |
Unconjugated | |
PDSS2 | |
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57107 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction