Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDSS2 Mouse anti-Human, Clone: UMAB177, lyophilized, UltraMAB™
SDP

Catalog No. 501685038
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5038 Supplier OriGene Supplier No. UM500108CF
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.

Specifications

Antigen PDSS2
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone UMAB177
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene PDSS2
Gene Accession No. Q86YH6
Gene Alias C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1
Gene Symbols PDSS2
Host Species Mouse
Immunogen Full length human recombit protein of human PDSS2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57107
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.