Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDSS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118153
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118153 Supplier Invitrogen™ Supplier No. PA5118153
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDSS2 Polyclonal Antibody for Western Blot, IHC (P), IP

The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDSS2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene PDSS2
Gene Accession No. Q86YH6
Gene Alias 5430420P03Rik; All-trans-decaprenyl-diphosphate synthase subunit 2; bA59I9.3; C6orf210; Candidate tumor suppressor protein; COQ10D3; decaprenyl diphosphate synthase subunit 2; decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; decaprenyl-diphosphate synthase subunit 2; DLP1; EGK_15212; Gm60; hDLP1; hypothetical protein LOC436624; kd; mDLP1; Pdss2; Plmp; prenyl (decaprenyl) diphosphate synthase, subunit 2; prenyl (solanesyl) diphosphate synthase, subunit 2; prenyl diphosphate synthase, subunit 2; prenyltransferase-like mitochondrial protein; RP11-59I9.3; solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase; zgc:92156
Gene Symbols PDSS2
Host Species Rabbit
Immunogen E. coli-derived Human PDSS2 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57107
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.