Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX-1/IPF1 Antibody (5A5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237506
Description
PDX-1/IPF1 Monoclonal specifically detects PDX-1/IPF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
PDX-1/IPF1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
Mouse | |
30.8 kDa | |
0.1 mL | |
Lipid and Metabolism, Stem Cell Markers | |
3651 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
5A5 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
PDX1 | |
Purified recombinant fragment of human PDX1 expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction