Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX-1/IPF1 Antibody (OTI2A12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273330F
Description
PDX-1/IPF1 Monoclonal specifically detects PDX-1/IPF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PDX-1/IPF1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PDX1 | |
Full length recombinant protein of human PDX-1/IPF1 (NP_000200) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2A12 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism, Stem Cell Markers | |
3651.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction