Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX-1/IPF1 Antibody (PCRP-PDX1-2C11), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308853MFV610
Description
PDX-1/IPF1 Monoclonal antibody specifically detects PDX-1/IPF1 in Human samples. It is validated for Flow CytometrySpecifications
PDX-1/IPF1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-PDX1-2C11 | |
Flow Cytometry | |
Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
Recombinant full-length human PDX-1/IPF1 protein (Uniprot: P52945) | |
0.1 mL | |
Lipid and Metabolism, Stem Cell Markers | |
3651 | |
Store at 4C in the dark. | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction