Learn More
PDX-1 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 601590

Description
Positive control: human pancreas tissues Cellular location: Nuclear.
PDX-1 Polyclonal antibody specifically detects PDX-1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PDX-1 | |
Polyclonal | |
Unconjugated | |
PDX1 | |
DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1 | |
A synthetic peptide of AEPEE NKR TRT AYTR derived from PDX1 protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P52945 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3651 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.