Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDX1 Monoclonal Antibody (OTI1E1), TrueMAB™, OriGene
SDP

Catalog No. 501681487
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1487 Supplier OriGene Supplier No. TA807312
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

pdx-1 (pancreas duodenum homeobox-1, also known as IPF-1, IDX-1, and STF-1) is a transcription factor that has been shown to activate insulin, glucokinase, IAPP, and somatostatin gene transcription. Significant interest surrounds this proteins glucose-dependent regulation of insulin gene transcription. Additionally, pdx-1 has been studied for its role in pancreatic development which includes maturation and differentiation of pancreatic precursor cells. Mutations in the pdx-1 gene have been shown to cause pancreatic agenesis, maturity-onset diabetes of the young, and type II diabetes.

Specifications

Antigen PDX1
Applications Western Blot
Classification Monoclonal
Clone OTI1E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDX1
Gene Accession No. P52945
Gene Alias GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
Gene Symbols PDX1
Host Species Mouse
Immunogen Full length human recombit protein of human PDX1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3651
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.