Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX1 Monoclonal Antibody (OTI4H7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807341
Description
pdx-1 (pancreas duodenum homeobox-1, also known as IPF-1, IDX-1, and STF-1) is a transcription factor that has been shown to activate insulin, glucokinase, IAPP, and somatostatin gene transcription. Significant interest surrounds this proteins glucose-dependent regulation of insulin gene transcription. Additionally, pdx-1 has been studied for its role in pancreatic development which includes maturation and differentiation of pancreatic precursor cells. Mutations in the pdx-1 gene have been shown to cause pancreatic agenesis, maturity-onset diabetes of the young, and type II diabetes.Specifications
| PDX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52945 | |
| PDX1 | |
| Full length human recombit protein of human PDX1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4H7 | |
| Unconjugated | |
| PDX1 | |
| GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3651 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction