Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX1 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 61289
Description
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).Specifications
| PDX1 | |
| Polyclonal | |
| Unconjugated | |
| PDX1 | |
| BOS_12504; DLDBP; E3BP; Glucose-sensitive factor; GSF; homeobox protein PDX-1; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; Insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; Islet/duodenum homeobox-1; IUF1; IUF-1; LOW QUALITY PROTEIN: pancreas/duodenum homeobox protein 1; Mody4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic and duodenum homeobox 1; pancreatic duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; PDX-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; Somatostatin-transactivating factor 1; STF1; STF-1; zgc:100874 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3651 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Lot-Specific | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| P52945 | |
| PDX1 | |
| This PDX1 antibody was raised against a peptide within the N-terminal region of human PDX1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction