Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587695
Description
Immunogen sequence: MNGEEQYYAA TQLYKDPCAF QRGPAPEFSA SPPACLYMGR QPPPPPPHPF PGALGALEQG SPPDISPYEV PPLADDPAVA HLHHHLPAQL ALPHPPAGPF; Positive Samples: DU145, HepG2, Mouse pancreas, Mouse liver; Cellular Location: Cytoplasm, Nucleus, cytosol.
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).
Specifications
| PDX1 | |
| Polyclonal | |
| Unconjugated | |
| PDX1 | |
| DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18609, 3651 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52945, P52946 | |
| PDX1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PDX1 (NP_0002001). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction