Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDXDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550100 Supplier Invitrogen™ Supplier No. PA550100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PDXDC1 protein.

PDXDC1 (Pyridoxal Dependent Decarboxylase Domain Containing 1) is a Protein Coding gene. Diseases associated with PDXDC1 include Autosomal Recessive Nonsyndromic Deafness 32 and Fetal Akinesia Deformation Sequence 1. Gene Ontology (GO) annotations related to this gene include pyridoxal phosphate binding and carboxy-lyase activity. An important paralog of this gene is HDC. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDXDC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pdxdc1
Gene Accession No. Q6P996, Q99K01
Gene Alias 2210010A19Rik; AA415817; KIAA0251; Kiaa0251-hp; LP8165; Pdxdc1; pyridoxal dependent decarboxylase domain containing 1; pyridoxal-dependent decarboxylase domain containing 1; pyridoxal-dependent decarboxylase domain-containing protein 1; RGD1562597
Gene Symbols Pdxdc1
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human PDXDC1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23042, 304721, 94184
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.