Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDXDC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568945
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8945 Supplier Bethyl Laboratories, Inc Supplier No. A304888A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Belongs to the group II decarboxylase family. 2 isoforms of the human protein are produced by alternative splicing. Nagase T., DNA Res. 3:321-329(1996). The MGC Project Team; Genome Res. 14:2121-2127(2004). Li H., Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases.

Specifications

Antigen PDXDC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PDXDC1
Gene Accession No. Q6P996
Gene Alias LP8165, pyridoxal-dependent decarboxylase domain-containing protein 1
Gene Symbols PDXDC1
Host Species Rabbit
Immunogen Between 550 and 600
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23042
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.