Learn More
PDXDC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304890AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Belongs to the group II decarboxylase family. 2 isoforms of the human protein are produced by alternative splicing. Nagase T., DNA Res. 3:321-329(1996). The MGC Project Team; Genome Res. 14:2121-2127(2004). Li H., Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases.Specifications
PDXDC1 | |
Polyclonal | |
Unconjugated | |
PDXDC1 | |
LP8165, pyridoxal-dependent decarboxylase domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23042 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6P996 | |
PDXDC1 | |
Between 738 and 788 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.