Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDXK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598044 Supplier Thermo Scientific Supplier No. PA598044
Add to Cart
Add to Cart

PDXK Polyclonal Antibody

PDXK Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDXK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDXK
Gene Accession No. O00764, Q8K183
Gene Alias 2310036D04Rik; AA119688; C21orf124; C21orf97; C77999; epididymis secretory sperm binding protein Li 1a; HEL-S-1a; Pdxk; Pkh; PNK; PRED79; pyridoxal (pyridoxine, vitamin B6) kinase; pyridoxal kinase; pyridoxamine kinase; pyridoxine kinase; vitamin B6 kinase
Gene Symbols PDXK
Host Species Rabbit
Immunogen Recombinant Human Pyridoxal kinase protein (1-312AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216134, 8566
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.