Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDXP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110095 Supplier Invitrogen™ Supplier No. PA5110095
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDXP Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP. Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP (Jang et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDXP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PDXP
Gene Accession No. P60487, Q8VD52, Q96GD0
Gene Alias 1600027H05Rik; AB041662; chronophilin; chronophin; CIN; dJ37E16.5; FLJ32703; PDXP; Plp; PLP phosphatase; PLPP; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; pyridoxal phosphatase; pyridoxal phosphate phosphatase; Rbp1; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36
Gene Symbols PDXP
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57026, 57028, 727679
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.