Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDXP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142628
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142628 Supplier Invitrogen™ Supplier No. PA5142628
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP. Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP (Jang et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDXP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PDXP
Gene Accession No. P60487, Q8VD52, Q96GD0
Gene Alias 1600027H05Rik; AB041662; chronophilin; chronophin; CIN; dJ37E16.5; FLJ32703; PDXP; Plp; PLP phosphatase; PLPP; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; pyridoxal phosphatase; pyridoxal phosphate phosphatase; Rbp1; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36
Gene Symbols PDXP
Host Species Goat
Immunogen Peptide with sequence C-QHDLVPHYYVES.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57026, 57028, 727679
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.