Learn More
PDZD11 Antibody, DyLight 594, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP299951DL594
Catalog No. NB063643
Only null left
Add to Cart
Description
Description
PDZD11 Polyclonal specifically detects PDZD11 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
PDZD11
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
AIPP1, ATPase-interacting PDZ protein, PDZ domain containing 11, PDZ domain-containing protein 11, PDZK11, PISP
Produced in rabbits immunized with purified, recombinant PDZD11 (NP_057568.1; Asp 2-His 140).
0.1 mL
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 594
50mM Sodium Borate
Rabbit
Affinity purified
RUO
51248
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.