Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547800
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547800 Supplier Invitrogen™ Supplier No. PA547800
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 5% cross-reactivity with recombinant human PDZD2 and recombinant mouse PDZD2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZD11
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PDZD11
Gene Accession No. Q5EBL8
Gene Alias AIPP1; ATPase-interacting PDZ protein; HSPC227; PDZ domain containing 11; PDZ domain-containing protein 11; PDZD11; PDZK11; PISP; Plasma membrane calcium ATPase-interacting single-PDZ protein; PMCA-interacting single-PDZ protein; UNQ6486/PRO21335
Gene Symbols PDZD11
Host Species Goat
Immunogen E. coli-derived recombinant human PDZD11 Met1-His140.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51248
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.