Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552033 Supplier Invitrogen™ Supplier No. PA552033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPYDDYPVVF LPAYENPPAW IPPHERVHHP DYNNELTQFL PRTITLKKPP GAQLGFNIRG GKASQLGIFI SKVIPDSDAH RAGLQEGDQV LAVNDVDFQD IEHSKAVEIL KTAREISMRV RFFPYNYHR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZD11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDZD11
Gene Accession No. Q5EBL8
Gene Alias AIPP1; ATPase-interacting PDZ protein; HSPC227; PDZ domain containing 11; PDZ domain-containing protein 11; PDZD11; PDZK11; PISP; Plasma membrane calcium ATPase-interacting single-PDZ protein; PMCA-interacting single-PDZ protein; UNQ6486/PRO21335
Gene Symbols PDZD11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDZD11. Recombinant protein control fragment (Product #RP-92331).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.