Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDZD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546583
Description
Peptide sequence: PDPWSLERPR FCLLSKEEGK SFGFHLQQEL GRAGHVVCRV DPGTSAQRQG Sequence homology: Cow: 79%; Dog: 93%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 86%; Rat: 93%.
Acts as a regulatory protein that associates with GUCY2C and negatively modulates its heat-stable enterotoxin-mediated activation.
Specifications
PDZD3 | |
Polyclonal | |
Unconjugated | |
Pdzd3 | |
DLNB27; hypothetical protein LOC539171; IKEPP; intestinal and kidney enriched PDZ protein; intestinal and kidney-enriched PDZ protein; Na(+)/H(+) exchange regulatory cofactor NHE-RF4; na/Pi cotransporter C-terminal-associated protein 2; NaPi-Cap2; natrium-phosphate cotransporter IIa C-terminal-associated protein 2; Nherf4; NHERF-4; PDZ domain containing 2; PDZ domain containing 3; PDZ domain-containing protein 2; PDZ domain-containing protein 3; Pdzd3; Pdzk2; RGD1559807; Sodium-hydrogen exchanger regulatory factor 4; sodium-phosphate cotransporter IIa C-terminal-associated protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79849 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86UT5 | |
Pdzd3 | |
Synthetic peptide directed towards the N-terminal of human PDZD3 (aa 105-154). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction