Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140211 Supplier Invitrogen™ Supplier No. PA5140211
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PSTLSGHRVC QAHGEPVLGL CPLLPLFCCP PHPPDPWSLE RPRFCLLSKE EGKSFGFHLQ QELGRAGHVV CRVDPGTSAQ RQGLQEGDRI LAVNNDV Highest antigen sequence indentity to the following orthologs: Mouse - 55%, Rat - 55%.

Acts as a regulatory protein that associates with GUCY2C and negatively modulates its heat-stable enterotoxin-mediated activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pdzd3
Gene Accession No. Q86UT5
Gene Alias DLNB27; hypothetical protein LOC539171; IKEPP; intestinal and kidney enriched PDZ protein; intestinal and kidney-enriched PDZ protein; Na(+)/H(+) exchange regulatory cofactor NHE-RF4; na/Pi cotransporter C-terminal-associated protein 2; NaPi-Cap2; natrium-phosphate cotransporter IIa C-terminal-associated protein 2; Nherf4; NHERF-4; PDZ domain containing 2; PDZ domain containing 3; PDZ domain-containing protein 2; PDZ domain-containing protein 3; Pdzd3; Pdzk2; RGD1559807; Sodium-hydrogen exchanger regulatory factor 4; sodium-phosphate cotransporter IIa C-terminal-associated protein 2
Gene Symbols Pdzd3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDZD3. Recombinant protein control fragment (Product #RP-109656).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.