Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDZD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570930
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
The function of this protein remains unknown.
Specifications
PDZD4 | |
Polyclonal | |
Unconjugated | |
PDZD4 | |
B230341P03; D130075J17Rik; KIAA1444; Kiaa1444-hp; LNX5; LU1; PDZ domain containing 4; PDZ domain-containing protein 4; PDZ domain-containing RING finger protein 4-like protein; PDZD4; Pdzk4; PDZRN4L; Pdzrnk4l; RGD1561214; Xlu | |
Rabbit | |
Affinity chromatography | |
RUO | |
245469 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9QY39 | |
PDZD4 | |
synthetic peptide directed towards the C-terminal region of mouse PDZD4. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction