Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDZD4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571832
Description
The function of this protein remains unknown.
Specifications
PDZD4 | |
Polyclonal | |
Unconjugated | |
PDZD4 | |
PDZ domain-containing protein 4; PDZ domain-containing RING finger protein 4-like protein; PDZD4; KIAA1444; PDZK4; PDZRN4L | |
Rabbit | |
Protein A | |
RUO | |
245469, 57595 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q76G19, Q9QY39 | |
PDZD4 | |
KLH conjugated synthetic peptide between 113-141 amino acids from the N-terminal region of human PDZD4 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction