Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584076 Supplier Invitrogen™ Supplier No. PA584076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEEHIHIQQW ALTEGRLKVT LLECSRLLIF GSYDREANVH CTLELSSSVW EEKQRSSIKT VELIKGNLQS VGLTLRLVQS TDGYAGHVII ETVAPNSPAA IADLQRGDR Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 96%.

PDZD8 plays a role in the regulation of cell morphology and cytoskeletal organization. It may inhibit herpes simplex virus 1 infection at an early stage and may promote retroviral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZD8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDZD8
Gene Accession No. Q8NEN9
Gene Alias A630041P07Rik; AV329468; AW228944; bA129M16.2; FLJ34427; PDZ domain containing 8; PDZ domain-containing protein 8; Pdzd8; PDZK8; Sarcoma antigen NY-SAR-84/NY-SAR-104
Gene Symbols PDZD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDZD8. Recombinant protein control fragment (Product #RP-106348).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.