Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDZK1 Antibody (SR2158), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PDZK1 |
---|---|
Clone | SR2158 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
PDZK1 Monoclonal antibody specifically detects PDZK1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PDZK1 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAP70, CFTR-associated protein of 70 kDa, CLAMP, Na(+)/H(+) exchanger regulatory factor 3, Na/Pi cotransporter C-terminal-associated protein 1, naPi-Cap1, NHERF-3, NHERF3PDZD1Na(+)/H(+) exchange regulatory cofactor NHE-RF3, PDZ domain containing 1, PDZ domain-containing protein 1, PDZ-containing kidney protein 1, Sodium-hydrogen exchanger regulatory factor 3 | |
A synthesized peptide derived from human PDZK1 (Uniprot #: Q5T2W1) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2158 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cellular Markers, Diabetes Research, Endothelial Cell Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Plasma Membrane Markers, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5174 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title