Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDZK1, Mouse anti-Human, Clone: AT1A2, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16352
Specifications
PDZK1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (10% glycerol, 0.02% sodium azide). | |
CAP70/CLAMP/NHERF3/PDZD1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
AT1A2 | |
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
PDZ domain containing 1 | |
PDZK1 | |
Recombinant protein corresponding to amino acids 1-519 of human PDZK1 from E. coli. | |
RUO | |
5174 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction