Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZK1 Polyclonal Antibody

Catalog No. PIPA552199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552199 Supplier Invitrogen™ Supplier No. PA552199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDASHEEVVE KVKKSGSRVM FLLVDKETDK RHVEQKIQFK RETASLKLLP HQPRIVEMKK GSNGYGFYLR AGSEQKGQII KDIDSGSPAE EAGLKNNDLV VAVNGESVET Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

PDZK1 is a scaffold protein that connects plasma membrane proteins and regulatory components, regulating their surface expression in epithelial cells apical domains. It may be involved in the coordination of a diverse range of regulatory processes for ion transport and second messenger cascades. In complex with SLC9A3R1, it may cluster proteins that are functionally dependent in a mutual fashion and modulate the trafficking and the activity of the associated membrane proteins. It may also play a role in the cellular mechanisms associated with multidrug resistance through its interaction with ABCC2 and PDZK1IP1. May potentiate the CFTR chloride channel activity. PDZK1 functions to connect SCARB1 with the cellular machineries for intracellular cholesterol transport and/or metabolism and may be involved in the regulation of proximal tubular Na(+)-dependent inorganic phosphate cotransport therefore playing an important role in tubule function.

Specifications

Antigen PDZK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDZK1
Gene Accession No. Q5T2W1
Gene Alias 1700023D20Rik; 2610507N21Rik; 4921513F16Rik; AI267131; AI314638; AL022680; CAP70; CFTR-associated protein of 70 kDa; Clamp; C-terminal linking and modulating protein; C-terminal-linking and modulating protein; D3Ertd537e; Dietary Pi-regulated RNA-1; Diphor1; diphor-1; mPDZK1; Na(+)/H(+) exchange regulatory cofactor NHE-RF3; Na(+)/H(+) exchanger regulatory factor 3; Na/Pi cotransporter C-terminal-associated protein 1; NaPi-C; naPi-Cap1; NHERF3; NHERF-3; PDZ domain containing 1; PDZ domain-containing protein 1; PDZ-containing kidney protein 1; Pdzd1; PDZK 1; Pdzk1; PDZK-1; Sodium-hydrogen exchanger regulatory factor 3
Gene Symbols PDZK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDZK1. Recombinant protein control fragment (Product #RP-88793).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5174
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.