Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDZK1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9036R

Description
PDZK1 is a scaffold protein that connects plasma membrane proteins and regulatory components, regulating their surface expression in epithelial cells apical domains. It may be involved in the coordination of a diverse range of regulatory processes for ion transport and second messenger cascades. In complex with SLC9A3R1, it may cluster proteins that are functionally dependent in a mutual fashion and modulate the trafficking and the activity of the associated membrane proteins. It may also play a role in the cellular mechanisms associated with multidrug resistance through its interaction with ABCC2 and PDZK1IP1. May potentiate the CFTR chloride channel activity. PDZK1 functions to connect SCARB1 with the cellular machineries for intracellular cholesterol transport and/or metabolism and may be involved in the regulation of proximal tubular Na(+)-dependent inorganic phosphate cotransport therefore playing an important role in tubule function.Specifications
PDZK1 | |
Polyclonal | |
Unconjugated | |
PDZK1 | |
1700023D20Rik; 2610507N21Rik; 4921513F16Rik; AI267131; AI314638; AL022680; BOS_3043; Cap70; CFTR-associated protein of 70 kDa; Clamp; C-terminal linking and modulating protein; C-terminal-linking and modulating protein; D3Ertd537e; Dietary Pi-regulated RNA-1; Diphor1; Diphor-1; mPDZK1; Na(+)/H(+) exchange regulatory cofactor NHE-RF3; Na(+)/H(+) exchanger regulatory factor 3; Na/Pi cotransporter C-terminal-associated protein 1; NaPi-C; NaPi-Cap1; NHERF3; NHERF-3; PDZ domain containing 1; PDZ domain-containing protein 1; PDZ-containing kidney protein 1; PDZD1; PDZK 1; Pdzk1; PDZK-1; Sodium-hydrogen exchanger regulatory factor 3 | |
Rabbit | |
Protein A | |
RUO | |
5174 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5T2W1 | |
PDZK1 | |
KLH conjugated synthetic peptide derived from human PDZK1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction