Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDZRN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558553
Description
Immunogen sequence: PDNSLRRAAE GISCPSSEGA VGTTEAYGPA SKNLLSITED PEVGTPTYSP SLKELDPNQP LESKERRASD GSRSPTPSQK LGSAYLPSYH HSP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
E3 ubiquitin-protein ligase. Plays an important role in regulating the surface level of MUSK on myotubes. Mediates the ubiquitination of MUSK, promoting its endocytosis and lysosomal degradation. Might contribute to terminal myogenic differentiation.
Specifications
PDZRN3 | |
Polyclonal | |
Unconjugated | |
PDZRN3 | |
1110020C07Rik; AI429718; AL023082; E3 ubiquitin-protein ligase PDZRN3; KIAA1095; Ligand of Numb protein X 3; likely ortholog of mouse semaF cytoplasmic domain associated protein 3; LNX3; PDZ domain containing ring finger 3; PDZ domain containing RING finger 3A; PDZ domain-containing RING finger protein 3; Pdzrn3; Protein SEMACAP3; RING-type E3 ubiquitin transferase PDZRN3; SEMACAP3; semaphorin cytoplasmic domain-associated protein 3; semaphorin cytoplasmic domain-associated protein 3A; Semcap3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23024 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPQ7 | |
PDZRN3 | |
Recombinant protein corresponding to Human PDZRN3. Recombinant protein control fragment (Product #RP-96326). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction