Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZRN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566390 Supplier Invitrogen™ Supplier No. PA566390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEQENNGDDAT ASSNPLAGQR KLTCSQDTLG SGDLPFSNES FISADCTDAD YLGIPVDECE RFR Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.

E3 ubiquitin-protein ligase. Plays an important role in regulating the surface level of MUSK on myotubes. Mediates the ubiquitination of MUSK, promoting its endocytosis and lysosomal degradation. Might contribute to terminal myogenic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZRN3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDZRN3
Gene Accession No. Q9UPQ7
Gene Alias 1110020C07Rik; AI429718; AL023082; E3 ubiquitin-protein ligase PDZRN3; KIAA1095; Ligand of Numb protein X 3; likely ortholog of mouse semaF cytoplasmic domain associated protein 3; LNX3; PDZ domain containing ring finger 3; PDZ domain containing RING finger 3A; PDZ domain-containing RING finger protein 3; Pdzrn3; Protein SEMACAP3; RING-type E3 ubiquitin transferase PDZRN3; SEMACAP3; semaphorin cytoplasmic domain-associated protein 3; semaphorin cytoplasmic domain-associated protein 3A; Semcap3
Gene Symbols PDZRN3
Host Species Rabbit
Immunogen Recombinant Human PDZRN3. Recombinant protein control fragment (Product #RP-107064).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.