Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZRN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583591 Supplier Invitrogen™ Supplier No. PA583591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WRRREKALLA QLWALQGEVQ LTARRYQEKF TQYMAHVRNF VGDLGGGHRR DGEHKPFTIV LERE Highest antigen sequence indentity to the following orthologs: Mouse - 48%, Rat - 45%.

PDZRN4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZRN4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDZRN4
Gene Accession No. Q6ZMN7
Gene Alias IMAGE5767589; ligand of Numb protein X 4; LNX4; PDZ domain containing ring finger 4; PDZ domain-containing RING finger protein 4; PDZRN4; SAMCAP3L; SEMACAP3-like protein; SEMCAP3L
Gene Symbols PDZRN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDZRN4. Recombinant protein control fragment (Product #RP-108193).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.