Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE-CF594 Mouse Anti-Human TACTILE (CD96), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570151
Description
TACTILE (CD96) Monoclonal antibody specifically detects TACTILE (CD96) in Human samples. It is validated for Flow cytometry
Specifications
| TACTILE (CD96) | |
| Monoclonal | |
| PE-CF594 | |
| hCD96; T cell-activated increased late expression protein | |
| Human NK92 Cell Line | |
| 25 Tests | |
| 10225 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry | |
| NK92.39.rMAb | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction