Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE-CF594 Mouse Anti-Human TCR Vβ13.1, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569816
Description
TCR Vβ13.1 Monoclonal antibody specifically detects TCR Vβ13.1 in Human samples. It is validated for Flow cytometry
Specifications
TCR Vβ13.1 | |
Monoclonal | |
PE-CF594 | |
TCR V beta 13.1; TCR Vbeta13.1; TCRBV13; TCRBV13.1; TCRBV13S1; TRBV6-1 | |
Human TCR Vβ13.2 Element Transfected Cell Line | |
100 Tests | |
28606 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2b κ |
Flow Cytometry | |
H131 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction