Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Human BCMA (CD269), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570547
Description
BCMA (CD269) Monoclonal antibody specifically detects BCMA (CD269) in Human samples. It is validated for Flow cytometry
Specifications
BCMA (CD269) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
19F2.rMAb | |
PE | |
B-cell maturation protein; BCM; TNFRSF17 | |
Human BCMA Ectodomain-Mouse IgG Fc | |
25 μg | |
608 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction