Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Human CD36, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570215
Description
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow cytometry
Specifications
CD36 | |
Monoclonal | |
PE | |
GPIIIb; Glycoprotein IIIb; PAS IV; Platelet glycoprotein IV; Scavenger receptor class B, member 3; Thrombospondin receptor | |
Fetal Erythrocytes | |
25 Tests | |
948 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
FA6-152 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction